Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Dihydroethidium ≥98% | Avantor

Dihydroethidium ≥98%

25 mg
Katalognr. CAYM12013-25
Anbieter:   Cayman Chemical
Dihydroethidium ≥98%
Katalognr. CAYM12013-25
Anbieter:  Cayman Chemical
CAS Nummer:  104821-25-2
Dihydroethidium ≥98%

25 mg

CAS Nummer:  104821-25-2
Preis
SFr. 394.00
Einloggen für Verfügbarkeit
1 * 25 mg
Product Details & Documents

Dihydroethidium is a cell-permeable blue fluorescent dye that upon entering cells interacts with superoxide to form oxyethidium, which intercalates with nucleic acids and emits a red fluorescence detectable qualitatively by fluorescent microscopy or quantitatively by HPLC.

It displays excitation/emission spectra of 490/590 nm. This selective and sensitive probe can be used to detect reactive oxygen species during the phagocytic respiratory burst and for the detection of intracellular superoxide in cultured cells.

Synonyme:  3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine
  • CAS Nummer:104821-25-2
  • Formel:C₂₁H₂₁N₃
  • MDL-Nummer:MFCD00077335
  • Molekulargewicht:315.412 g/mol
  • Lagertemperatur:Gefrierschrank
Technische Daten

Ergebnisse des Spezifikationstests

  • Purity:
    ≥98%
  • Emission:
    590 nm
  • Excitation:
    490 nm
  • Formulation:
    crystalline solid
  • λmax:
    224, 271, 354 nm

Technische Daten

  • VE:
    25 mg

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.