Digestion system, behrotest®
Complete crude fibre digestion unit with 4 sample positions, EXR 4
Katalognr. BEHRB00218446
Anbieter: BEHR
Direkt zu
Product Details & Documents
Digestion apparatus for crude fibre or hydrolysis.
Lieferinformationen: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Technische Daten
- Bezeichnung:Complete crude fibre digestion unit with 4 sample positions
- Seq:02
- Typ:EXR 4