Warning signs
Warning laser beam, PP, fest, 315×315 mm
Catalog # BRDY252219
Supplier: Brady
Jump to:
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Specifications
- Bezeichnung:Warning laser beam
- Abmessungen:315×315 mm
- Material:PP, fest
- Seq:32