Optical bottom plates, SCREENSTAR
Product Details & Documents
Cycloolefin, black pigmented frame, 190 μm ultra-clear film bottom, tissue culture treated, sterile. SCREENSTAR microplates are specialised products for sophisticated microscopic applications, like high content screening (HCS) or high resolution microscopy with water and oil immersion objectives.
- Resistant against polar solvents (DMSO)
- Glass-like optical qualities with a refractive index similar to glass
- Ideal surface for adherent cell culture
- Universal microscope objective compatibility
Manufactured entirely of cycloolefin, the microplates feature a black pigmented frame with a 190 μm ultra-clear film bottom for ideal compatibility with instrument optics. Well bottoms display excellent optical properties for the highest optical transparency with reduced autofluorescence in the lower UV range, low birefringence and a refractive index of 1.53 comparable to glass. Recessed microplate wells enable complete periphery access for high magnification objectives.
Technische Daten
- Bezeichnung:96-well SCREENSTAR microplate, TC-treated, with lid
- Cat. No.:735-0387
- Umweltzertifizierung:ISO 14001 (241002); ISO 50001 (241002)
- Umweltfreundlichkeit:Umweltschonende Herstellung