AuftragsverfolgungKontakt
Cylindrical bend testers
Cylindrical bend testers
Cylindrical bend tester (100 mm) with desk holder and seven mandrels (Ø = 1/8 to 1") 
Catalog # SIMESP1822
Supplier:   Industrial Physics
Cylindrical bend testers
Catalog # SIMESP1822
Supplier:  Industrial Physics
Cylindrical bend testers
Cylindrical bend testers
Cylindrical bend tester (100 mm) with desk holder and seven mandrels (Ø = 1/8 to 1") 
Price
SFr. 2’940.00
Log in for availability
1 * 1 ST
Product Details & Documents
These cylindrical bend testers are used to determine the elasticity, adhesion and elongation of paint on metal.
  • Sturdy conception in anodised aluminium and stainless steel
  • Ergonomic clamping device for test panels
  • Large knob on bending arm for smooth bending
The test panel is bent over the cylindrical mandrel and then checked for damages.
Certifications: ISO 1519 (or ASTM D522)
Delivery information: Supplied with a desktop holder and mandrels.
Documents
Specifications
  • Bezeichnung:
    Cylindrical bend tester (100 mm) with desk holder and seven mandrels (Ø = 1/8 to 1") 
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1840
    Mandrel, 6 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1842
    Mandrel, 10 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1835
    Set of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holder
    Log in for availability
    1 * 14 ST - SFr. 690.00
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1836
    Mandrel, 2 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1837
    Mandrel, 3 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1838
    Mandrel, 4 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1839
    Mandrel, 5 mm
    Log in for availability
    1 * 1 ST - SFr. 75.90
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1843
    Mandrel, 12 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1841
    Mandrel, 8 mm
    Log in for availability
    1 * 1 ST - SFr. 75.90
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1844
    Mandrel, 13 mm
    Log in for availability
    1 * 1 ST - SFr. 75.90
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1845
    Mandrel, 16 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1846
    Mandrel, 19 mm
    Log in for availability
    1 * 1 ST - SFr. 75.90
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1847
    Mandrel, 20 mm
    Log in for availability
    1 * 1 ST - SFr. 88.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1848
    Mandrel, 25 mm
    Log in for availability
    1 * 1 ST - SFr. 75.90
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1849
    Mandrel, 32 mm
    Log in for availability
    1 * 1 ST - SFr. 75.90
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1833
    Set of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holder
    Log in for availability
    1 * 1 ST - SFr. 697.00
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1828
    Mandrel, 3/4"
    Log in for availability
    1 * 1 ST - SFr. 248.00
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1823
    Mandrel, 1/8"
    Log in for availability
    1 * 1 ST - SFr. 194.00
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1824
    Mandrel, 1/4"
    Log in for availability
    1 * 1 ST - SFr. 248.00
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1825
    Mandrel, 3/8"
    Log in for availability
    1 * 1 ST - SFr. 242.00
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1826
    Mandrel, 1/2"
    Log in for availability
    1 * 1 ST - SFr. 194.00
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • SIMESP1827
    Mandrel, 5/8"
    Log in for availability
    1 * 1 ST - SFr. 242.00