Warning signs
Warning radioactive substances, PE, 25×22 mm
Catalog # BRDY826758
Supplier: Brady
Jump to:
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Specifications
- Bezeichnung:Warning radioactive substances
- Abmessungen:25×22 mm
- Material:PE
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.