Combi top racks
30/20/12/10 mm, Blau, 4/12/32/32, 50/15/1,5/0,5 ml, 180×90×52 mm
Codice catalogo KOEHA-1183
Nome del fornitore: KOEHLER TECHNISCHE PRODUKTEN
Vai a
Product Details & Documents
Racks, made of polypropylene, neon coloured racks have four sides that can be used individually. with temperature resistant from −80 to 121 °C.
- Temperature resistant from −80 to 121 °C.
- Excellent chemical resistance
- Sturdy
Specifiche
- Für Röhrchen:50/15/1,5/0,5 ml
- Bohrung Ø:30/20/12/10 mm
- Anz. Bohrungen:4/12/32/32
- Farbe:Blau
- Seq:01
- B×T×H:180×90×52 mm