Okadasäure ≥98%
100 µG
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 9,10-deepithio-9,10-didehydroacanthifolicin, OA
- CAS Nummer:78111-17-8
- Formel:C₄₄H₆₈O₁₃
- MDL-Nummer:MFCD00083455
- Molekulargewicht:805.02 g/mol
- Merck-Index:14,6819
- Lagertemperatur:Gefrierschrank
- UN:3462
- ADR:6.1,I
Dokumente
Technische Daten
Technische Daten
- VE:100 µG
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



