Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Nitrogen generator, Precision nitrogen 600

Nitrogen generator, Precision nitrogen 600

Precision 600, 600 ml/min, 380×540×256 mm
N° de catalogue PEAK61-0600
Fournisseur:   PEAK SCIENTIFIC
Nitrogen generator, Precision nitrogen 600
N° de catalogue PEAK61-0600
Fournisseur:  PEAK SCIENTIFIC
Nitrogen generator, Precision nitrogen 600
Nitrogen generator, Precision nitrogen 600

Precision 600, 600 ml/min, 380×540×256 mm

Prix
SFr. 7’390.00
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

The Precision Nitrogen 600 has been developed to provide a constant and consistent source of nitrogen for detector make-up gas at standard/typical detection levels for GC applications, as well as headspace vial pressurisation, purge and trap, tube conditioning for thermal desorption and sample preparation. These generators are capable of delivering high purity nitrogen, removing oxygen and moisture via Pressure Swing Adsorption and Carbon Molecular Sieve technology.

  • Suitable as make-up gas at standard detection limits
  • Constant and consistent supply, avoid running out of gas during analysis
  • Dedicated precision compressor module available where house supply not available
  • Integrates seamlessly with other precision units
  • 1 year comprehensive on-site warranty

Nitrogen model has a compact and modular, stackable design, minimizing the total footprint required for GC gas supply, and providing flexibility to add or remove modules as the laboratory requirements evolve over time. Frequency: 50/60 Hz.

Inlet pressure: 6,8 to 8,2 bar

Outlet pressure: 5,5 bar

Weight: 21 kg

Current: 0,2 to 0,4 A

Certifications: Fully compliant with: 2012/19/EU WEEE (Waste of Electrical and Electronic Equipment) directive;
2011/65/EU RoHS 2 (Restriction of Hazardous Substance).

Spécifications
  • Förderleistung:
    600 ml/min
  • B×T×H:
    380×540×256 mm
  • Typ:
    Precision 600
  • Cat. No.:
    PEAK61-0600
  • Gewicht:
    41 kg