THE PALL™ VECTOR SYSTEM WAS DEVELOPED SO THAT FOLLOWING DIGESTION WITH NSI1 AN RNA CAN BE GENERATED USING SP6 RNA POLYMERASE. 1 * 10 µG
Katalognr. BIOO5205-03
Anbieter: BIOO SCIENTIFIC
THE PALL™ VECTOR SYSTEM WAS DEVELOPED SO THAT FOLLOWING DIGESTION WITH NSI1 AN RNA CAN BE GENERATED USING SP6 RNA POLYMERASE. 1 * 10 µG
Katalognr. BIOO5205-03
Anbieter: BIOO SCIENTIFIC
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Sealing film for manual application, Parafilm® M
Anbieter: HEATHROW
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
Anbieter: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
Anbieter: VWR Chemicals
VWR®, Serologische Pipetten, Standard Serie
Anbieter: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
Anbieter: KIMBERLY CLARK
VWR®, Nitrilhandschuhe
Anbieter: VWR Collection
VWR® Gesichtstücher
Anbieter: VWR Collection
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
Anbieter: VWR Chemicals
pH-Indikatorstäbchen, MQuant®, Supelco®
Anbieter: Merck
VWR® NITRILE LIGHT, Nitrilhandschuhe
Anbieter: VWR Collection
VWR®, Transferpipetten, High Performance
Anbieter: VWR Collection