96-Well Blocks
96-well block, 2 blocks, 2 ml
Katalognr. ZYMOP1001-2
Marketsource
Anbieter: Zymo Research
Direkt zu
Product Details & Documents
Deep-well block, 2 ml for processing and mixing of samples.
- Conforms to laboratory standards
96-well block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Technische Daten
- Verpackt:2 blocks
- Bezeichnung:96-well block
- Seq:01
- Gesamtvolumen:2 ml