AuftragsverfolgungKontakt
Zymo-Spin™ Column/Filter Assemblies | Avantor
Zymo-Spin™ Column/Filter Assemblies
ZRC-GF Filter™, Spin column/filter assembly, 20
Katalognr. ZYMOC1009-20
Marketsource
Anbieter:   Zymo Research
Zymo-Spin™ Column/Filter Assemblies
Katalognr. ZYMOC1009-20
Anbieter:  Zymo Research
Marketsource
CAS Nummer:  
Zymo-Spin™ Column/Filter Assemblies
Zymo-Spin™ Column/Filter Assemblies
ZRC-GF Filter™, Spin column/filter assembly, 20
CAS Nummer:  
Preis
SFr. 91.00
Einloggen für Verfügbarkeit
1 * 20 ST
Product Details & Documents
Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.
  • For purifying DNA and or/RNA from high-volume sample inputs
  • High-recovery
The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA.
The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.
The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1,6 μm pore size glass fibre filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.
Technische Daten
Technische Daten
  • VE:
    20
  • Bezeichnung:
    ZRC-GF Filter™
  • Format:
    Spin column/filter assembly
Technische Daten
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..