Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Zymo-Spin™ Column/Filter Assemblies | Avantor

Zymo-Spin™ Column/Filter Assemblies

Zymo-Spin™ VI with Zymo-Maxi Filter™, Spin column/filter assembly, 20
Katalognr. ZYMOC1017-20
Marketsource
Anbieter:   Zymo Research
Zymo-Spin™ Column/Filter Assemblies
Katalognr. ZYMOC1017-20
Anbieter:  Zymo Research
Marketsource
CAS Nummer:  
Zymo-Spin™ Column/Filter Assemblies
Zymo-Spin™ Column/Filter Assemblies

Zymo-Spin™ VI with Zymo-Maxi Filter™, Spin column/filter assembly, 20

CAS Nummer:  
Preis
SFr. 127.00
Einloggen für Verfügbarkeit
1 * 20 ST
Product Details & Documents

Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.

  • For purifying DNA and or/RNA from high-volume sample inputs
  • High-recovery

The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA.

The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.

The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.

The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1,6 μm pore size glass fibre filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.

Technische Daten

Technische Daten

  • VE:
    20
  • Bezeichnung:
    Zymo-Spin™ VI with Zymo-Maxi Filter™
  • Format:
    Spin column/filter assembly

Technische Daten