O-rings
O-rings, viton, type KF, DN 16
Catalog # ILMV701171
Supplier: Welch by Gardner Denver
Jump to:
Product Details & Documents
O-rings, Type KF made of Perbunan® or Viton®.
Specifications
- Bezeichnung:O-rings, viton, type KF, DN 16
- Seq:03
O-rings, Type KF made of Perbunan® or Viton®.