AuftragsverfolgungKontakt
Erlenmeyer flasks, narrow neck, with screw cap, Corning® | Avantor
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
100 ml, GL 40
Katalognr. CORN4990P-100
Anbieter:   Corning
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Katalognr. CORN4990P-100
Anbieter:  Corning
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
100 ml, GL 40
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 6 ST
Product Details & Documents
PMP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With easy to read red markings.
Technische Daten
  • Gewinde:
    GL 40
  • Kapazität:
    100 ml
  • Seq:
    02