Erlenmeyer flasks, narrow neck, with screw cap, Corning®
100 ml, GL 40
Katalognr. CORN4990P-100
Anbieter: Corning
Direkt zu
Product Details & Documents
PMP, translucent.
- Very good chemical resistance
- Very good temperature resistance
With easy to read red markings.
Technische Daten
- Gewinde:GL 40
- Kapazität:100 ml
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.