AuftragsverfolgungKontakt
Erlenmeyer flasks, narrow neck, with screw cap, Corning® | Avantor
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
1000 ml, GL 52
Katalognr. CORN4990P-1L
Anbieter:   Corning
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Katalognr. CORN4990P-1L
Anbieter:  Corning
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
1000 ml, GL 52
Preis
SFr. 119.00
Einloggen für Verfügbarkeit
1 * 4 ST
Product Details & Documents
PMP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With easy to read red markings.
Technische Daten
  • Gewinde:
    GL 52
  • Kapazität:
    1000 ml
  • Seq:
    05