Erlenmeyer flasks, narrow neck, with screw cap, Corning®
250 ml, GL 52
Catalog # CORN4990P-250
Supplier: Corning
Jump to:
Product Details & Documents
PMP, translucent.
- Very good chemical resistance
- Very good temperature resistance
With easy to read red markings.
Specifications
- Gewinde:GL 52
- Kapazität:250 ml
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.