AuftragsverfolgungKontakt
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
500 ml, GL 52
Catalog # CORN4990P-500
Supplier:   Corning
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Catalog # CORN4990P-500
Supplier:  Corning
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
500 ml, GL 52
Price
SFr. 141.00
Log in for availability
1 * 6 ST
Product Details & Documents
PMP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With easy to read red markings.
Specifications
  • Gewinde:
    GL 52
  • Kapazität:
    500 ml
  • Seq:
    04