Erlenmeyer flasks, narrow neck, with screw cap, Corning®
500 ml, GL 52
Catalog # CORN4990P-500
Supplier: Corning
Jump to:
Product Details & Documents
PMP, translucent.
- Very good chemical resistance
- Very good temperature resistance
With easy to read red markings.
Specifications
- Gewinde:GL 52
- Kapazität:500 ml
- Seq:04