15ml Nanosph.Size Stds 60nm 1 * 15 mL
MSPP3060A
Frequently Bought Together
Dekontaminationslösung RNase AWAY®
: Thermo Fisher Scientific
VWR®, Petrischalen, PS
: VWR Collection
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Transferpipetten, Standard Line
: VWR Collection
DNA AWAY® Dekontaminationslösung
: Thermo Fisher Scientific
VWR®, Transferpipetten, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
Einwegspritzen, dreiteilig, HENKE-JECT®
: HENKE SASS WOLF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF