Gowning bench
Gowning bench, L×W: 1825×406 mm, 31,8 kg
Catalog # METOGB1672S
Supplier: Metro International
Jump to:
Product Details & Documents
Gowning bench in stainless steel 304.
- Heavy-duty, solid top bench provides comfortable gowning fixture
- Stainless steel foot plates provide floor mount or free-standing option
Documents
METOGB1636S_TDS_EN
Datasheet
Specifications
- Bezeichnung:Gowning bench, L×W: 1825×406 mm
- Seq:04
- Gewicht:31,8 kg