Serotonin Hydrochlorid
Glass bottle for solids, 10 g
Katalognr. S21263.09
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 5-Hydroxytryptamine Hydrochloride
- CAS Nummer:153-98-0
- Formel:C₁₀H₁₃ClN₂O
- MDL-Nummer:MFCD00012686
- Molekulargewicht:212.68 g/mol
- UN:2811
- ADR:6.1,III
Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:10 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



