Serotonin Hydrochlorid
Glass bottle for solids, 1 kg
Katalognr. S21263.A1
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 5-Hydroxytryptamine Hydrochloride
- CAS Nummer:153-98-0
- Formel:C₁₀H₁₃ClN₂O
- MDL-Nummer:MFCD00012686
- Molekulargewicht:212.68 g/mol
- UN:2811
- ADR:6.1,III
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 kg



