AuftragsverfolgungKontakt
Axygen® AxyPrep® Mag DyeClean kit
AxyPrep® Mag DyeClean Kit, 5 ml, 1 KIT
Katalognr. MAG-DYECL-5
Anbieter:   Corning
Axygen® AxyPrep® Mag DyeClean kit
Katalognr. MAG-DYECL-5
Anbieter:  Corning
CAS Nummer:  
Axygen® AxyPrep® Mag DyeClean kit
Axygen® AxyPrep® Mag DyeClean kit
AxyPrep® Mag DyeClean Kit, 5 ml, 1 KIT
CAS Nummer:  
Preis
SFr. 365.00
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Removing excess dye terminator is an essential step prior to Sanger sequencing. Carryover of the excess dye into the sequencing reactions may result in dye blobs resulting in inaccurate results. The AxyPrep™ Mag DyeClean kit utilises a paramagnetic bead-based purification system for Sanger sequencing reaction dye terminator clean-up. The protocol is simple and comprises binding, washing and elution steps which can be performed directly in the thermal cycling plate. This kit requires no centrifugation or filtration steps.
  • High signal to noise ratio resulting in long reads and high QV20+ scores
  • Suitable for the clean up of sequencing reaction of many different Sanger sequencing based platforms
  • Simple and easy sequencing reaction clean up process: 25 minutes/96 samples
  • Manual and automation friendly
  • Scalable: Tube, 96-well and 384-well plate formats
  • No centrifugation or filtration required
Technische Daten
Technische Daten
  • VE:
    1 KIT
  • Bezeichnung:
    AxyPrep® Mag DyeClean Kit, 5 ml
Technische Daten