Material pouch, snikki
Material pouch, leather, Braun
Katalognr. KWIN100014240
Anbieter: FRISTADS KANSAS
Direkt zu
Product Details & Documents
Pouch are made of heavy duty split leather that consist of two pockets for screws and nails with sheath knife holder, rivet reinforced and easy quick-grip.
Big pocket: 160×80×200 mm (W×D×H)
Small pocket: 140×45×130 mm (W×D×H)
Dokumente
Technische Daten
- Farbe:Braun
- Bezeichnung:Material pouch, leather
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



