2-Fluor-4-nitrophenol
Glasflasche für Feststoffe, 100 g
Catalog # ACTIAS20908G100
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Fluor-4-nitrophenol
Catalog # ACTIAS20908G100
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 403-19-0
Jump to:
Product Details & Documents
- CAS Number:403-19-0
- Formula:C₆H₄FNO₃
- MDL Number:MFCD00051970
- Molecular Weight:157.1 g/mol
- Melting Pt:118…118,5 °C
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



