4-Isopropylanilin
Glasflasche für Feststoffe, 25 g
ACTIAS21354G25
Marketsource
: p-Cumidine, 4-Aminocumene
- :99-88-7
- :C₉H₁₃N
- :MFCD00007900
- :135.21 g/mol
- :13,02647
- :226 °C (1013 hPa)
- :–63 °C
- :0,9526 g/cm³ (20 °C)
- :92 °C
- :Raumtemperatur
- :202-797-2
- :1760
- :8,III
- Packung:Glasflasche für Feststoffe
- VE:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.