4-Isopropylanilin
Glasflasche für Feststoffe, 100 g
Catalog # ACTIAS21354G100
Marketsource
Supplier: ACTIVATE SCIENTIFIC
4-Isopropylanilin
Catalog # ACTIAS21354G100
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 99-88-7
Jump to:
Product Details & Documents
Synonyms: p-Cumidine, 4-Aminocumene
- CAS Number:99-88-7
- Formula:C₉H₁₃N
- MDL Number:MFCD00007900
- Molecular Weight:135.21 g/mol
- Merck Index:13,02647
- Boiling Pt:226 °C (1013 hPa)
- Melting Pt:–63 °C
- Density:0,9526 g/cm³ (20 °C)
- Flash Pt:92 °C
- Storage Temperature:Raumtemperatur
- EINECS:202-797-2
- UN:1760
- ADR:8,III
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



