2'-(Trifluormethyl)acetophenon
Glass bottle for solids, 100 g
Katalognr. ACTIAS20270G100
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
2'-(Trifluormethyl)acetophenon
Katalognr. ACTIAS20270G100
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 17408-14-9
Direkt zu
Product Details & Documents
Synonyme: o-(Trifluoromethyl)acetophenone, 1-[2-(Trifluoromethyl)phenyl]ethan-1-one, 1-(2-(Trifluoromethyl)phenyl)ethanone
- CAS Nummer:17408-14-9
- Formel:C₉H₇F₃O
- MDL-Nummer:MFCD00000378
- Molekulargewicht:188.15 g/mol
- Siedepunkt:163 °C (1013 hPa)
- Schmelzender Platingehalt:15…17 °C
- Dichte:1,25 g/cm³ (20 °C)
- :Density: 1.25 g/cm³ (20 °C)
- EINECS:241-434-2
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



