4-Bromchinolin
Glasflasche für Feststoffe, 1 g
Katalognr. ACTIAS1343G1
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
4-Bromchinolin
Katalognr. ACTIAS1343G1
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 3964-04-3
Direkt zu
Product Details & Documents
- CAS Nummer:3964-04-3
- Formel:C₉H₆BrN
- MDL-Nummer:MFCD07644514
- Molekulargewicht:208.06
- Lagertemperatur:Kühlschrank
- UN:3261
- ADR:8,II
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



