3-Chlorisochinolin
Glass bottle for solids, 5 g
Katalognr. ACTIAS11198G5
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
3-Chlorisochinolin
Katalognr. ACTIAS11198G5
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 19493-45-9
Direkt zu
Product Details & Documents
- CAS Nummer:19493-45-9
- Formel:C₉H₆ClN
- MDL-Nummer:MFCD05982009
- Molekulargewicht:163.6000 g/mol
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



