3-Bromisochinolin
Glass bottle for solids, 1 g
Katalognr. ACTIAS1287G1
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
3-Bromisochinolin
Katalognr. ACTIAS1287G1
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 34784-02-6
Direkt zu
Product Details & Documents
- CAS Nummer:34784-02-6
- Formel:C₉H₆BrN
- MDL-Nummer:MFCD00234479
- Molekulargewicht:208.0600 g/mol
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



