2-(Difluormethoxy)phenylessigsäure
Glass bottle for solids, 1 g
Katalognr. ACTIAS34369G1
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
2-(Difluormethoxy)phenylessigsäure
Katalognr. ACTIAS34369G1
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 86867-68-7
Direkt zu
Product Details & Documents
- CAS Nummer:86867-68-7
- Formel:C₉H₈F₂O₃
- MDL-Nummer:MFCD00236237
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



