3-Fluor-2-nitrobenzonitril
Glasflasche für Feststoffe, 1 g
Katalognr. ACTIAS32653G1
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
3-Fluor-2-nitrobenzonitril
Katalognr. ACTIAS32653G1
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 1000339-52-5
Direkt zu
Product Details & Documents
- CAS Nummer:1000339-52-5
- Formel:C₇H₃FN₂O₂
- MDL-Nummer:MFCD09864664
- Molekulargewicht:166.1100 g/mol
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



