4-Methoxyanthranilsäure
Glass bottle for solids, 1 g
Katalognr. ACTIAS31288G1
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
4-Methoxyanthranilsäure
Katalognr. ACTIAS31288G1
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 4294-95-5
Direkt zu
Product Details & Documents
Synonyme: 2-Amino-4-methoxybenzoic acid
- CAS Nummer:4294-95-5
- Formel:C₈H₉NO₃
- MDL-Nummer:MFCD00667729
- Molekulargewicht:167.16 g/mol
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



