4-Phenylanilin
Glass bottle for solids, 1 g
Catalog # ACTIAS31139G1
Marketsource
Supplier: ACTIVATE SCIENTIFIC
Jump to:
Product Details & Documents
Synonyms: 4-Biphenylylamine, Biphenyl-4-ylamine, 4-Aminobiphenyl
- CAS Number:92-67-1
- Formula:C₁₂H₁₁N
- MDL Number:MFCD00007879
- Molecular Weight:169.23 g/mol
- Merck Index:12,01276
- Boiling Pt:302 °C (1013 hPa)
- Melting Pt:55 °C
- Density:1,16 g/cm³ (20 °C)
- :Dichte: 1,16 g/cm³ (20 °C)
- EINECS:202-177-1
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



