2-Methylbenzophenon
Glass bottle for solids, 25 g
ACTIAS25303G25
Marketsource
: o-Methylbenzophenone
- :131-58-8
- :C₁₄H₁₂O
- :MFCD00008518
- :196.25 g/mol
- :12,07470
- :310 °C (1013 hPa)
- :28 °C
- :1,1098 g/cm³ (20 °C)
- :205-032-0
- Packung:Glass bottle for solids
- VE:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.