Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Razor blades, double edge prep | Avantor

Razor blades, double edge prep

Double edge prep razor blades, individually wrapped, 0,10 mm, 38×19 mm
Katalognr. PERS74-0010-0000
Anbieter:   PERSONNA MEDICAL
Razor blades, double edge prep
Katalognr. PERS74-0010-0000
Anbieter:  PERSONNA MEDICAL
Razor blades, double edge prep

Double edge prep razor blades, individually wrapped, 0,10 mm, 38×19 mm

Preis
SFr. 184.00
Einloggen für Verfügbarkeit
1 * 250 ST
Product Details & Documents

Personna® Double Edge Prep razor blades are produced using innovative technologies. The high quality stainless steel is carefully selected for optimal charateristics and corrosion resistance.

  • Corrosion resistant
  • Optimised coating and geometry for superior performance
  • Reduced cut force
  • Blades individually wrapped

Dokumente

Technische Daten
  • Bezeichnung:
    Double edge prep razor blades, individually wrapped
  • L×B:
    38×19 mm
  • Seq:
    02
  • Stärke:
    0,10 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.