Microplates, 96-well, non binding
96-well microplate, V-bottom, non binding, solid bottom, Klar
Katalognr. 738-0166
Anbieter: Greiner Bio-One
Direkt zu
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Technische Daten
- Farbe:Klar
- Bezeichnung:96-well microplate, V-bottom, non binding, solid bottom
- Seq:02
- Cat. No.:738-0166
- Umweltzertifizierung:ISO 14001 (241002); ISO 50001 (241002)
- Umweltfreundlichkeit:Umweltschonende Herstellung