Phosphate, 0,2 M buffer solution, pH 7,5
Phosphate buffer soln, 0,2 M, 7,5, 1 L
Anbieter: Thermo Fisher Scientific
Phosphate, 0,2 M buffer solution, pH 7,5
Katalognr. J61324.K2
Anbieter: Thermo Fisher Scientific
CAS Nummer:
Direkt zu
Product Details & Documents
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Dokumente
Technische Daten
Technische Daten
- Konzentration:0,2 M
- VE:1 L
- Bezeichnung:Phosphate buffer soln
- pH-Wert:7,5
- Cat. No.:J61324.K2
- CAS Nr.:7558-80-7
- Physical form:Liquid
- MDL No.:MFCD00003527
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



