Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
L-thyroxine 98% | Avantor

L-thyroxine 98%

1 g
L-thyroxine 98%
Katalognr. J62606.03
CAS Nummer:  51-48-9
L-thyroxine 98%
L-thyroxine 98%

1 g

CAS Nummer:  51-48-9
Preis
SFr. 176.00
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 51-48-9 Molecular Formula: C15H11I4NO4 Formula Weight: 776.87 Physical Form: Crystalline powder Sensitivity: Light Sensitive Appearance: White to pale brown Odour: Odourless Melting Point: 235-236° Solubility: Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml Beilstein Reference: 2228515 MDL No.: MFCD00002595

Synonyme:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Nummer:51-48-9
  • Formel:C₁₅H₁₁I₄NO₄
  • MDL-Nummer:MFCD00002595
  • Molekulargewicht:776.874 g/mol
  • Merck-Index:14,9415
  • Schmelzender Platingehalt:235…236 °C
  • Lagertemperatur:Raumtemperatur
  • EINECS:200-101-1

Dokumente

Technische Daten

Technische Daten

  • VE:
    1 g

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.