Okadasäure ≥98%
300 µG
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA
- CAS Number:78111-17-8
- Formula:C₄₄H₆₈O₁₃
- MDL Number:MFCD00083455
- Molecular Weight:805.02 g/mol
- Merck Index:14,6819
- Storage Temperature:Gefrierschrank
- UN:3462
- ADR:6.1,I
Specifications
Specifications
- VE:300 µG



