TE buffer, (20X), pH 8,0, autoclaved
TE buffer concentrate, autoclaved, 20X, 8,0, 500 mL
Supplier: Thermo Fisher Scientific
TE buffer, (20X), pH 8,0, autoclaved
Catalog # J62388.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00236359
Documents
Specifications
Specifications
- Konzentration:20X
- VE:500 mL
- Bezeichnung:TE buffer concentrate, autoclaved
- pH-Wert:8,0
- Cat. No.:J62388.AP
- Physical form:Liquid
- MDL No.:MFCD00236359
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



