Natriumalginat
Glasflasche für Feststoffe, 2,5 kg
Katalognr. SPCMSO106-2.5KG
Anbieter: Spectrum Chemical
Direkt zu
Product Details & Documents
Synonyme: Alginic acid sodium salt, Algin
- CAS Nummer:9005-38-3
- MDL-Nummer:MFCD00081310
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:2,5 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



