AuftragsverfolgungKontakt
Stirrer blades | Avantor
Stirrer blades
Anchor stirrer blade, PTFE, with stiffened metal core, for SVL reaction vessels, 90 mm
Katalognr. BIBB588-95
Anbieter:   SciLabware
Stirrer blades
Katalognr. BIBB588-95
Anbieter:  SciLabware
Stirrer blades
Stirrer blades
Anchor stirrer blade, PTFE, with stiffened metal core, for SVL reaction vessels, 90 mm
Preis
153.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
A range of PTFE coated stirrer heads designed to screw into the threads on SVL® glass stirrer shafts.
  • Head simply screws into the end of the shaft
  • Anchor and helix types have a PTFE coated metal core
Achtung! Bitte beachten: Stirrers must be rotated in a clockwise direction to prevent unscrewing of the thread.
Technische Daten
  • Typ:
    Anchor stirrer blade, PTFE, with stiffened metal core, for SVL reaction vessels
  • Breite:
    90 mm
  • Seq:
    010
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..