Stirrer blades
Collapsible stirrer blade, PTFE, for vessels with SVL 30 joint or 29/32 socket, 70 mm
Katalognr. BIBB588-93
Anbieter: SciLabware
Direkt zu
Product Details & Documents
A range of PTFE coated stirrer heads designed to screw into the threads on SVL® glass stirrer shafts.
- Head simply screws into the end of the shaft
- Anchor and helix types have a PTFE coated metal core
Caution: Stirrers must be rotated in a clockwise direction to prevent unscrewing of the thread.
Technische Daten
- Typ:Collapsible stirrer blade, PTFE, for vessels with SVL 30 joint or 29/32 socket
- Breite:70 mm
- Seq:007
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.