AuftragsverfolgungKontakt
Stirrer blades | Avantor
Stirrer blades
Collapsible stirrer blade, PTFE, for vessels with SVL 30 joint or 29/32 socket, 70 mm
Katalognr. BIBB588-93
Anbieter:   SciLabware
Stirrer blades
Katalognr. BIBB588-93
Anbieter:  SciLabware
Stirrer blades
Stirrer blades
Collapsible stirrer blade, PTFE, for vessels with SVL 30 joint or 29/32 socket, 70 mm
Preis
SFr. 121.00
Einloggen für Verfügbarkeit
1 * 1 pezzi
Product Details & Documents
A range of PTFE coated stirrer heads designed to screw into the threads on SVL® glass stirrer shafts.
  • Head simply screws into the end of the shaft
  • Anchor and helix types have a PTFE coated metal core
Caution: Stirrers must be rotated in a clockwise direction to prevent unscrewing of the thread.
Technische Daten
  • Typ:
    Collapsible stirrer blade, PTFE, for vessels with SVL 30 joint or 29/32 socket
  • Breite:
    70 mm
  • Seq:
    007