Automatic burettes, with intermediate stopcock, class AS, Schellbach
25 ml, 0,05 ml, 0,03 ± ml
Katalognr. BIBB3496/04AS
Anbieter: SciLabware
Automatic burettes, with intermediate stopcock, class AS, Schellbach
Katalognr. BIBB3496/04AS
Anbieter: SciLabware
Direkt zu
Product Details & Documents
Burettes made of chemically resistant Pyrex® borosilicate glass and feature a 29/32 Quickfit® cone.
- With automatic zero facility
- Blue enamel graduations with Schellbach stripe
- PTFE stopcock
Schellbach burette tube with vertical white stripe and central blue ribbon, facilitates accurate reading of the meniscus, also exact position of the meniscus is clearly defined where the blue ribbon splits into a wide and narrow band.
Technische Daten
- Kapazität:25 ml
- Seq:02
- Teilung:0,05 ml
- Toleranz:0,03 ± ml