AuftragsverfolgungKontakt
Hardness tablets | Avantor
Hardness tablets
Hardness Yes/No tablets, AF423 Minikit, 250 Tab.
Katalognr. LOVI515361BT
Anbieter:   Lovibond
Hardness tablets
Katalognr. LOVI515361BT
Anbieter:  Lovibond
CAS Nummer:  
Hardness tablets
Hardness Yes/No tablets, AF423 Minikit, 250 Tab.
CAS Nummer:  
Preis
SFr. 126.00
Einloggen für Verfügbarkeit
1 * 250 Tablet
Product Details & Documents
The speed test is based on reverse titration. After adding a reagent tablet to a calibrated test tube, the water sample is added slowly until the colour of the solution changes (e.g. from red to blue). The user can then obtain the result from the liquid level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Dokumente
Technische Daten
Technische Daten
  • VE:
    250 Tab.
  • Bezeichnung:
    Hardness Yes/No tablets
  • Für Gerät:
    AF423 Minikit