Packaging Bags
0,09 mm, 500×700 mm
Catalog # SCAC3044
Supplier: SCANCELL FOLIEN VETRIEB
Jump to:
Product Details & Documents
These vacuum bags are with sealed edge and ideal for packaging of gas-sensitive goods and foods.
- PA/PE, transparent
Specifications
- Abmessungen:500×700 mm
- Seq:05
- Stärke:0,09 mm