n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
N° de catalogue 8088.9025
Fournisseur: Avantor
Numéro CAS: 110-54-3
Aller à
Product Details & Documents
Synonymes: Hexyl hydride, Hexanen, Hexane
- Numéro CAS:110-54-3
- Formule:H₃C(CH₂)₄CH₃
- Numéro MDL:MFCD00009520
- Poids moléculaire:86.18 g/mol
- Index Merck:13,04712
- Point d'ébullition:69 °C (1013 hPa)
- Point de fusion:–94,3 °C
- Densité:0,659 g/cm³ (20 °C)
- Point d'éclair:–22 °C
- Température de stockage:Raumtemperatur
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Spécifications
Résultats des tests de spécification
- Assay (by GC):min. 99 %
- Aromatic Compounds (as C6H6):max. 0,02 %
- Boiling Range:max. 2°C
- Density (g/ml) at 25°C:max. 0,657
- Recorded Boiling Point:68.7°C
- Residue after Evaporation:max. 5 ppm
- Sulfur Compounds (as S):max. 0,005 %
- Thiophene:passes test
- Water (H2O):max. 0,02 %
- Water Soluble Titrable Acid (meq/g):max. 0,0003
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Spécifications
- VE:25 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



