n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Glasflasche, 1 L
Anbieter: Avantor
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Katalognr. 8088.1000
Anbieter: Avantor
CAS Nummer: 110-54-3
Direkt zu
Product Details & Documents
Synonyme: Hexyl hydride, Hexanen, Hexane
- CAS Nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL-Nummer:MFCD00009520
- Molekulargewicht:86.18 g/mol
- Merck-Index:13,04712
- Siedepunkt:69 °C (1013 hPa)
- Schmelzender Platingehalt:–94,3 °C
- Dichte:0,659 g/cm³ (20 °C)
- Blitz Pt:–22 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Dokumente
Zertifikate
Suche nach Chargennummersds_BAKR8088 SDS_DE_CH_BAKR8088
Sicherheitsdatenblatt (SDS)
spec_BAKR8088.1000.pdf spec_BAKR8088.2500.pdf
Produktspezifikation
Technische Daten
Ergebnisse des Spezifikationstests
- Assay (by GC):min. 99 %
- Aromatic Compounds (as C6H6):max. 0,02 %
- Boiling Range:max. 2°C
- Density (g/ml) at 25°C:max. 0,657
- Recorded Boiling Point:68.7°C
- Residue after Evaporation:max. 5 ppm
- Sulfur Compounds (as S):max. 0,005 %
- Thiophene:passes test
- Water (H2O):max. 0,02 %
- Water Soluble Titrable Acid (meq/g):max. 0,0003
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Technische Daten
- Packung:Glasflasche
- VE:1 L



